Polypropylene (PP) Centrifuge Bottles: Translucent
Actual product may vary
Polypropylene (PP) Centrifuge Bottles: Translucent
Translucentt flat-bottom centrifuge bottles come with a high performance silicone O-ring cap or standard screw cap, depending on model
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Includes
FIS13-701-104250 mLSilicone O-ring cap
FIS13-701-105250 mLScrew cap
Catalog number FIS13-701-104
Price (SGD)
-
Capacity (Metric):
250 mL
Includes:
Silicone O-ring cap
Two easy grip cap styles: standard and high performance with silicone O-Ring
Note: Caps are not interchangeable among different materials or sizes.
  • Wide-mouth design improves liquid handling.
  • Compatible with most centrifuges and do not require special buckets
  • Extra-thick walls for durability
  • Leak-free processing at maximum rated speeds
  • Polypropylene bottles are autoclavable at 121°C, 15 psi for 20 minutes.
Specifications
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
ColorTranslucent
IncludesSilicone O-ring cap
MaterialPolypropylene
No. per Case36/Cs.
No. per Pack4/Pk.
SterilityNon-sterile
Closure ColorOrange
Graduations25 to 250 mL
GraduatedYes
Max. Force27,500 x g
MouthWide
ShapeCylindrical
Closure MaterialPolypropylene
Closure TypeO-Ring Cap