Internally Threaded Cryogenic Vials with Color Caps
Internally Threaded Cryogenic Vials with Color Caps
Actual product may vary
Internally Threaded Cryogenic Vials with Color Caps
Used for ultra-low temperature storage of valuable cell cultures, biological solutions and aqueous solutions
Have Questions?
Catalog NumberClosure Color
FIS13-700-500Assorted
Catalog number FIS13-700-500
Price (SGD)
1,385.00
Case
Add to cart
Closure Color:
Assorted
Price (SGD)
1,385.00
Case
Add to cart
  • Manufactured from polypropylene to withstand temperatures down to -196°C
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal.
  • 2 mL self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts.
  • Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Warning! Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen. Only store vials in the vapor phase above the liquefied gas. Always use appropriate safety equipment when removing vials from cryogenic storage.
Specifications
Closure ColorAssorted
ColorNatural
MaterialPolypropylene
SterilitySterile
Volume (Metric)2 mL
For Use With (Application)For ultra low temperature storage of valuable cell cultures, biological solutions and aqueous solutions
GraduatedYes
IncludesSilicone washer, inserts
No. per Case500
No. per Pack50
StyleRound
Temperature Range (English)-320.8°F
Temperature Range (Metric)-196°C
Thread StyleInternal
Base StyleRound (Self-standing)
RNase FreeYes
Dnase FreeYes
Closure MaterialPolypropylene
Marking SpotYes
Unit SizeCase