Square Polycarbonate Storage Bottles with 45 mm Cap
Actual product may vary
Square Polycarbonate Storage Bottles with 45 mm Cap
Space-Saving square shape requires less space (13 to 20%) on the shelf or in the autoclave. Corning™ Square Polycarbonate Storage Bottles are ideal for mixing, sampling and storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS13-700-412500 mL
FIS13-700-400150 mL
FKS10692434150 mL
FIS13-700-401250 mL
Catalog number FIS13-700-412
Price (SGD)
845.00
Case
Capacity (Metric):
500 mL
Price (SGD)
845.00
Case
Corning™ square polycarbonate storage bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage.
Caution: These square polycarbonate bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.
  • Strong polycarbonate bottles are more break resistant than other glass or plastic bottles.
  • Use a linerless, 45mm one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
  • Screened white enamel graduations are easier to see than molded graduations
  • Sterilized by gamma radiation
  • Large white marking spot for easier identification
  • Bottles can be autoclaved once at 121oC and 15psi. Repeated autoclaving weakens polycarbonate and is not recommended.
  • Optional reusable caps with silicone septa (Corning Cat. Nos. 1395-45HTSC, 1395-45SS, 1395-45TS) for syringe sampling or introduction of reagents are available for these caps.
  • Note: Most GL45 threaded caps designed for glass storage bottles use a different sealing design and will not give a secure seal if used on these plastic bottles unless they have a rubber liner.
  • Specifications
    Capacity15 oz., 500 mL
    Capacity (Metric)500 mL
    Capacity (English)15 oz.
    MaterialPolycarbonate
    No. per Case24/Cs.
    No. per Pack12/Pk.
    SterilitySterile
    Closure ColorOrange
    Graduations50 to 150 mL
    ShapeSquare
    Closure MaterialPolypropylene
    Closure SizeGL 45
    Closure TypeScrew Cap