BLAUBRAND™ Glass Graduated Cylinders, Class A, USP, Certified
Actual product may vary
BrandTech
BLAUBRAND™ Glass Graduated Cylinders, Class A, USP, Certified
Class A, USP Certified, calibrated 'To Contain'
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS13-690-152100 mL1 mL
FIS13-690-15150 mL1 mL
Catalog number FIS13-690-152
Price (SGD)
191.00
Pack
Capacity (Metric):
100 mL
Graduations:
1 mL
Price (SGD)
191.00
Pack
  • Manufactured from Borosilicate 3.3 glass, known for its low expansion coefficient and excellent chemical resistance
  • Features a blue enamel print for optimal durability and easy readability
  • Equipped with pour spout and a sturdy hexagonal glass base for convenient handling and stability
  • Available in a full range of ISO sizes from 5 mL to 2000 mL
  • Calibrated 'To Contain'
  • Each unit is supplied with batch number and a USP batch certificate in original packaging for traceability
Specifications
MaterialBorosilicate Glass
Capacity (Metric)100 mL
IncludesUSP Batch Certificate
Graduations1 mL
Subdivision1 mL
CalibrationTo Contain
Base TypeHexagonal
Scale ColorBlue
ShapeRound
ClassClass A
Height (Metric)260 mm
Certifications/ComplianceDIN EN ISO 4788, USP
Tolerance±0.5 mL
Product TypeGraduated Cylinder
Unit SizePack