Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Pipet Products
▸
Volumetric Pipets
▸
BLAUBRAND™ One-mark Glass Bulb Pipets, Class A, USP, Certified
Actual product may vary
BrandTech
BLAUBRAND™ One-mark Glass Bulb Pipets, Class A, USP, Certified
High contrast blue enamel represents optimum combination of resistance and readability
Have Questions?
Change view
Catalog Number
Volume (Metric)
FIS13-690-105
0.5 mL
FIS13-690-120
40 mL
2 Options
Catalog number
FIS13-690-105
Price (SGD)
495.00
Pack
Add to cart
Volume (Metric):
0.5 mL
Price (SGD)
495.00
Pack
Add to cart
Made of AR-GLAS™
Optimized flow geometry in tip ensures reliable liquid dispensing from pipet
Fire-polished edges for more durability
Available in all ISO sizes from 0.5 to 100mL
Supplied with batch number and an accompanying USP batch certificate in original packaging
Specifications
Certifications/Compliance
USP
Class
A
Material
AR-GLAS
Sterility
Non-sterile
Graduated
No
Color
Black
Volume (Metric)
0.5 mL
Length (Metric)
300 mm
Type
Bulb Pipet
Disposable
Reusable
For Use With (Equipment)
Pipet Controller