Macro Pipet Controller
Actual product may vary
BrandTech
Macro Pipet Controller
Firmly holds and fills all volumetric and serologic pipets from 0.10 to 200mL. Macro Pipet Controller provides better airflow for effortless bellows compression. A single pipetting lever controls filling, meniscus setting, and discharge.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FIS13-688-853Green
FIS13-688-850Gray
Catalog number FIS13-688-853
Price (SGD)
395.00
Each
Color:
Green
Price (SGD)
395.00
Each
Firmly holds and fills all volumetric and serologic pipets from 0.10 to 200mL. BrandTech™ Macro Pipet Controller provides better airflow for effortless bellows compression. A single pipetting lever controls filling, meniscus setting, and discharge. Built-in, replaceable 3μm hydrophobic filter (spare included) limits over-aspiration of liquids into the valve system. Autoclavable at 121°C.
Ergonomic pipette controller features a unique valve system that provides better airflow for effortless bellows compression.
  • Firmly holds and fills all volumetric and serologic pipets from 0.10 to 200mL
  • A single pipetting lever controls filling, meniscus setting, and discharge
  • Convenient blow-out bulb above the lever permits accurate dispensing from blow-out pipets
  • Easy-to-use controls and light weight (106g) reduces stress of repetitive pipetting
  • Built-in, replaceable 3μm hydrophobic filter (spare included) limits over-aspiration of liquids into the valve system
  • Silicone pipet adapter is replaceable
  • Entire instrument is autoclavable at 121°C for 20 minutes
Specifications
Adjustable Tip SpacingNo
AutoclavableAutoclavable
ColorGreen
DisposableReusable
For Use WithSerologic Pipette
Grip TypeBulb
MaterialPolypropylene, Silicone Rubber
Number of Channels1
SterilityNon-sterile
TypePipet Controller
Volume (Metric)0.1 to 200 mL
For Use With (Equipment)Serologic Pipettes
Quantity1 Each
Unit SizeEach