Transferpettor™ Pipetters
Actual product may vary
BrandTech
Transferpettor™ Pipetters
Handle difficult-to-pipet media such as viscous, foaming, or high vapor pressure liquids with precision and accuracy. Transferpettor™ Pipetters use tightly fitted piston with seal in direct contact with the aspirated liquid so results are reproducible. Walls are wiped clean to the last drop, and tips can be used repeatedly.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorIncrements
FIS13-688-252500 μLGreen1μL
FIS13-688-12110 mLOrange0.01mL
Catalog number FIS13-688-252
Price (SGD)
1,559.00
Each
Volume (Metric):
500 μL
Color:
Green
Increments:
1μL
Price (SGD)
1,559.00
Each
Handle difficult-to-pipet media such as viscous, foaming, or high vapor pressure liquids with precision and accuracy. Liquids must be compatible with polypropylene and high-density polyethylene.Four color-coded models cover range from 100μL to 10mL.
  • Tightly fitted piston with seal is in direct contact with the aspirated liquid so results are reproducible
  • Walls are wiped clean to the last drop, and tips can be used repeatedly
  • Accuracy: ≤±0.5%
  • Reproducibility: ≤0.2%
  • Maximum Vapor Pressure: 500 millibar
  • Maximum Viscosity: 20,000 cSt (mm2/sec)
  • Maximum Density: 13.6 g/cm3
Specifications
Accuracy≤±0.5%
Compatible TipsTransferpettor Tips, 100-500 μL
AutoclavableNot Autoclavable
ColorGreen
MaterialPolypropylene
Number of Channels1
TypePositive Displacement Pipette
Volume (Metric)500 μL
For Use With (Application)Liquids such as concentrated protein solutions, Oils, Fats, Resins, Glycerin high vapor pressure media, such as Alcohol, Acetone and Hydrocarbons
IncludesOne tip and one seal
Increments1μL
Repeatability±0.5% or less
Warranty1 Year
Unit SizeEach