Silicone Hand Protector
Actual product may vary
Silicone Hand Protector
Gripping surface is studded with small nubs for protection and sure handling of objects with temperatures ranging from -50°C to 260°C.
Have Questions?
Catalog number FIS13-684-147
Price (SGD)
19.00
Each
Price (SGD)
19.00
Each
  • Ideal for handling extremely hot or cold objects, including dry ice
  • Provides protection from temperatures ranging from -50°C to 260°C (-70°F to 500°F)
  • Silicone rubber conforms to the hand and object being handled
  • Gripping surface studded with small nubs for positive grip and temperature protection
  • One size fits all hands, left or right
  • Tab for hanging near work station
Specifications
ColorBlue
HandAmbidextrous
Latex FreeYes
MaterialSilicone Rubber
SizeUniversal
Temperature (English) Max.500°F
Temperature (Metric) Max260°C
For Use With (Application)Handling Extreme Temperature Objects
Temperature (English) Min-70°F
Temperature (Metric) Min.-50°C
Length (English)7.5 in.
Length (Metric)19 cm
Unit SizeEach