Sodium chloride, ultra dry, 99.998% (metals basis)
Sodium chloride, ultra dry, 99.998% (metals basis)
Sodium chloride, ultra dry, 99.998% (metals basis)
DWK Life Sciences
Sodium chloride, ultra dry, 99.998% (metals basis)
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Catalog NumberQuantity
FIS13-683-1425 g
Catalog number FIS13-683-14
Price (SGD)
1,415.00
Case
Quantity:
25 g
Request bulk or custom format
Price (SGD)
1,415.00
Case
Specifications
Boiling Point1413°C
Chemical Name or MaterialSodium chloride
Melting Point801°C
Name NoteUltra dry
CAS7647-14-5
View more
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media, a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only