Disposable Controlled-Drop Pasteur Pipets
Actual product may vary
Fisherbrand
Disposable Controlled-Drop Pasteur Pipets
Perfect for blood typing and cross-matching procedures in blood bank laboratories
Have Questions?
Catalog NumberMaterial
FIS13-678-30Borosilicate Glass
Catalog number FIS13-678-30
Price (SGD)
242.00
Case
Material:
Borosilicate Glass
Price (SGD)
242.00
Case
Perfect for blood typing and cross-matching procedures in blood bank laboratories
  • Deliver 22 to 26 drops per mL
  • Correspond to drop size from antisera reagent bottle droppers
  • Can be cotton plugged (plugs not included)
Specifications
Diameter (Metric) InnerTip Opening: 1.20 to 1.65 mm
Diameter (Metric) Outer6.95 mm (top)
MaterialBorosilicate Glass
For Use With (Application)For blood typing and cross-matching procedures in blood bank laboratories
Length (English)5.75 in.
Volume (Metric)2 mL
Length (Metric)14.6 cm
TypeControlled Drop Pasteur Pipet