Check Order Status
Reusable Cotton-Plug Top Serological Pipets
Actual product may vary
Reusable Cotton-Plug Top Serological Pipets
Provide fast, uniform flow times.
Have Questions?
Change view
Catalog Number
Volume (Metric)
Graduations
Accuracy
Packaging Type
FIS13-675M
10 mL
0.1 mL
±0.06 mL
Standard Pack
FIS13-675K
5 mL
0.1 mL
±0.04 mL
Standard Pack
2 Options
Catalog number
FIS13-675M
Price (SGD)
403.00
CS
Add to cart
Volume (Metric):
10 mL
Graduations:
0.1 mL
Accuracy:
±0.06 mL
Packaging Type:
Standard Pack
Price (SGD)
403.00
CS
Add to cart
Provide fast, uniform flow times.
Constriction in glass top prevents cotton plug from going down into pipet (plugs not included)
Accuracy printed on each pipet
Borosilicate glass with permanent amber markings
Tuf-Temp™ treated to reduce chipping
Specifications
Accuracy
±0.06 mL
Aspirator Plug
Yes
Certifications/Compliance
ASTM E1044-96
Material
Borosilicate Glass
Graduated
Yes
Graduations
0.1 mL
Packaging Type
Standard Pack
Volume (English)
0.33 oz.
Color
Yellow
Volume (Metric)
10 mL
Type
Serological Pipet