Standard-Top Serological Pipets
Standard-Top Serological Pipets
Actual product may vary
Standard-Top Serological Pipets
Provide fast, uniform flow times.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)AccuracyColor
FIS13-675H2 mL±0.02 mLGreen
FIS13-675F1 mL±0.02 mLYellow
Catalog number FIS13-675H
Price (SGD)
324.00
CS
Add to cart
Volume (Metric):
2 mL
Accuracy:
±0.02 mL
Color:
Green
Price (SGD)
324.00
CS
Add to cart
Provide fast, uniform flow times.
Accuracy printed on each pipet
  • Borosilicate glass, permanent amber markings
  • Tuf-Temp™ treated to reduce chipping
  • Pipets 13-675D and 13-675E are recommended for VDRL test
Specifications
Accuracy±0.02 mL
Certifications/ComplianceASTM E1044-96
MaterialBorosilicate Glass
GraduatedYes
Graduations0.1 mL
Packaging TypeStandard Pack
Volume (English)0.06 oz.
ColorGreen
Volume (Metric)2 mL
TypeSerological Pipet
Unit SizeCS