Polystyrene Aspirating Pipets, Without Graduations
Polystyrene Aspirating Pipets, Without Graduations
Actual product may vary
Polystyrene Aspirating Pipets, Without Graduations
Manufactured from polystyrene as a safer alternative to glass Pasteur pipets. Falcon™ Disposable Aspirating Pipets are ideal for tissue culture usage or any non-toxic solution that requires aliquoted or aspirating/dispensing in a sterile environment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FIS13-675-10CC5 mL
CRG3575582 mL
FIS13-675-162 mL
Catalog number FIS13-675-10CC
Price (SGD)
434.00
CS
Add to cart
Volume (Metric):
5 mL
Price (SGD)
434.00
CS
Add to cart
  • Sterile, polystyrene, non-plugged, non-graduated pipets
  • Safer alternative to glass Pasteur pipets
  • Can be used for all vacuum-aspirating procedures
  • Noncytotoxic, nonpyrogenic tested to less than 0.1 EU/mL
  • Individually packaged in thermoformed paper-plastic wrap to ensure sterile presentation
Specifications
Certifications/ComplianceClass I Medical
DescriptionAspirating
MaterialPolystyrene
SterilitySterile
For Use With (Application)For tissue culture usage or any non-toxic solution that requires aliquot or aspirating, dispensing in a sterile environment
Length (English)13.71 in.
Volume (Metric)5 mL
TypeAspirating Pipet