Measuring (Mohr) Standard-Taper Pipets
Measuring (Mohr) Standard-Taper Pipets
Actual product may vary
Measuring (Mohr) Standard-Taper Pipets
Accuracy is printed on each pipet.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)GraduationsColorAccuracy
FIS13-665N25 mL0.1 mLWhite±0.1 mL
FIS13-665F1 mL0.01 mLYellow±0.02 mL
FIS13-665G1 mL0.1 mLRed±0.02 mL
FIS13-665H2 mL0.1 mLGreen±0.02 mL
FIS13-665K5 mL0.1 mLBlue±0.04 mL
FIS13-665M10 mL0.1 mLOrange±0.06 mL
Catalog number FIS13-665N
Price (SGD)
447.00
CS
Add to cart
Volume (Metric):
25 mL
Graduations:
0.1 mL
Color:
White
Accuracy:
±0.1 mL
Price (SGD)
447.00
CS
Add to cart
  • Borosilicate glass
  • Permanent amber markings
  • Fast uniform flow
  • Tuf-Temp™-treated to reduce chipping
Specifications
Accuracy±0.1 mL
Certifications/ComplianceNNN-P-350C, ASTM E1293-94
MaterialBorosilicate Glass
GraduatedYes
Graduations0.1 mL
IncludesPermanent Amber marking
Volume (English)0.84 oz.
ColorWhite
Volume (Metric)25 mL
TypeMeasuring Standard Taper Pipet