Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets
Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™-51 Brand Reusable Class B Mohr (Measuring) Pipets
Rugged construction
Have Questions?
Catalog NumberVolume (Metric)ColorTolerance
FIS13-664H25 mLWhite±0.1 mL
Catalog number FIS13-664H
Price (SGD)
940.00
EA
Add to cart
Volume (Metric):
25 mL
Color:
White
Tolerance:
±0.1 mL
Price (SGD)
940.00
EA
Add to cart
Rugged construction
  • Intended for chemical laboratory work—tip openings are smaller than usually desired for clinical laboratory operations
  • Permanent brown, graduated scale fused into uniform bore tubing ending above taper
  • Color coded by size for ease in sorting
  • Tempered tips
  • Calibrated “To Deliver”
Specifications
Certifications/ComplianceASTM E438, USP Type I, Class B
ClassB
MaterialBorosilicate Glass
GraduatedYes
Graduations0.1 mL
Length (English)15.7 in.
Tolerance±0.1 mL
Volume (English)0.84 oz.
ColorWhite
Volume (Metric)25 mL
Length (Metric)400 mm
TypeGraduated Pipet, Type 1