Reusable Class B Volumetric Pipets
Reusable Class B Volumetric Pipets
Actual product may vary
Reusable Class B Volumetric Pipets
Ensure the highest accuracy and reproducibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Min. Flow TimeAccuracy
FIS13-650U100 mL20 sec.±0.16 mL
FIS13-650C2 mL3 sec.±0.012 mL
FIS13-650F5 mL8 sec.±0.02 mL
FIS13-650N20 mL10 sec.±0.06 mL
FIS13-650P25 mL15 sec.±0.06 mL
FIS13-650S50 mL15 sec.±0.1 mL
FIS13-650L10 mL8 sec.±0.04 mL
Catalog number FIS13-650U
Price (SGD)
77.00
EA
Add to cart
Volume (Metric):
100 mL
Min. Flow Time:
20 sec.
Accuracy:
±0.16 mL
Price (SGD)
77.00
EA
Add to cart
Ensure the highest accuracy and reproducibility.
  • Borosilicate glass
  • Permanent amber markings
  • Large numeral on bulb of 15mL and larger pipets makes capacity easy to identify
  • Accuracy printed on each pipet
  • Tuf-Temp™–treated tempered tips reduce chipping
  • Fast outflow
Specifications
Accuracy±0.16 mL
CalibrationTo Deliver
Certifications/ComplianceASTM E969-95
ClassB
MaterialBorosilicate Glass
GraduatedYes
Graduation ColorAmber
Min. Flow Time20 sec.
Tip StyleTuf-Temp™ treated
ColorYellow
Volume (Metric)100 mL
TypeVolumetric Pipet