Orion™ Redox/ORP/Temp Electrodes
Actual product may vary
Thermo Scientific
Orion™ Redox/ORP/Temp Electrodes
Accurately measure ORP and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Catalog NumberTypeFor Use With (Equipment)
FIS13-642-255Epoxy Triode Low MaintenanceOrion Star Series Meters
Catalog number FIS13-642-255
Price (SGD)
1,774.00
Each
Type:
Epoxy Triode Low Maintenance
For Use With (Equipment):
Orion Star Series Meters
Price (SGD)
1,774.00
Each
Thermo Scientific™ Orion™ Triode 3-in-1 pH/Automatic Temperature Compensation Electrodes come with a built-in ATC and are engineered with an epoxy body for ruggedness. The 9179BN and 9179BNMD are low-maintenance gel-filled electrodes, best suited for educational institutes where minimal maintenance is required, while the 9180BNMD is a refillable electrode best suited for general purpose samples.
Orion™ Single Junction Electrodes provide a reliable option for routine ORP measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required:
Electrode fill solution for 9180BNMD
  • 900011 ORP filling solution for samples with total ionic strength > 0.2 M, 5 x 60 mL bottles
  • 900001 ORP filling solution for samples with total ionic strength < 0.2 M, 5 x 60 mL bottles
Electrode storage solution
  • 910001 4 M KCl storage solution, 475 mL bottle
Standards
  • 967901 ORP standard, 475 mL bottle
  • 967961 ORP standard, 5 x 60 mL bottles
pH cleaning solution kit
  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each
Items included with 9810BNMD
  • 900011 ORP filling solution for samples with total ionic strength > 0.2 M, 60 mL 1 bottle
Warranty period
  • 9179BN and 9179BNMD: 6 months from date of purchase
  • 9180BNMD: 12 months from date of purchase
Specifications
ApplicationsWater testing,wastewater, disinfection, aquaculture, process water.
Connector TypesBNC (Waterproof)
DescriptionEpoxy Triode Low Maintenance
MaterialEpoxy
Certifications/ComplianceASTM D1498, Standard Methods 2580, USGS Ch 6.5
For Use With (Equipment)Orion Star Series Meters
Temperature (Metric) Operating0°C to 80°C
TypeEpoxy Triode Low Maintenance
Unit SizeEach