Orion™ ISE Filling Solutions
Actual product may vary
Thermo Scientific
Orion™ ISE Filling Solutions
Enhance the performance and extend the life of your ion selective electrodes with these ISE filling solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFor Use With (Equipment)
FIS13-641-8465 x 60 mLSodium ISE
FIS13-642-6325 x 60 mLCadmium ISE, Calcium ISE, Fluoride ISE, Sulfide ISE
FKS120545025 x 60 mLCadmium ISE, Calcium ISE, Fluoride ISE, Sulfide ISE
9000625 x 60 mLChloride ISE, Cyanide ISE, Lead ISE, Silver/Sulfide ISE
FIS13-641-1615 x 60 mLBromide ISE, Cupric ISE, Iodide ISE
FIS13-641-76660 mLCarbon Dioxide ISE
FKS1171767760 mLCarbon Dioxide ISE
FIS13-641-92860 mLAmmonia ISE
Catalog number FIS13-641-846
Price (SGD)
613.00
Pack
Quantity:
5 x 60 mL
For Use With (Equipment):
Sodium ISE
Price (SGD)
613.00
Pack
Enhance the performance and extend the life of your ion selective electrodes with Thermo Scientific™ Orion™ ISE Filling Solutions. These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.
Features:
  • Filling/maintenance solutions are for use with Thermo Scientific™ Orion™ Ion Selective Electrodes (ISE)
  • Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective electrodes
Ordering Information:
  • Packaged as set of five 2 oz. (5 x 60mL) bottles, except Cat. No. 951209, 951202 and 950202 packaged as one 2 oz. (60mL) bottle and Cat. No. 954602 packaged as one 1.6 oz. (50mL) bottle
Note:
The following part number has been changed due to a recent system upgrade:
Old Part Number: 900018
New Part Number: 900018-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFill Solution for Sodium ISE
For Use With (Equipment)Sodium ISE
TypeISE Filling Solution
Quantity5 x 60 mL
Unit SizePack