Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Actual product may vary
Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Fisherbrand
Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector Types
FIS13-641-780BNC (Waterproof), 8-pin Mini-DIN
FKS12493617BNC (Waterproof), 8-pin Mini-DIN
Catalog number FIS13-641-780
Price (SGD)
2,682.00
Each
Connector Types:
BNC (Waterproof), 8-pin Mini-DIN
Price (SGD)
2,682.00
Each
Thermo Scientific™ Refillable Orion™ ROSS Ultra™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra™ line with a built-in ATC. Best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. Refillable Orion™ ROSS Ultra™ Triodes™ come with a two year warranty.
Orion™ ROSS Ultra™ Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonymous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
24 months from date of purchase
Specifications
DescriptionOrion ROSS Ultra Glass Triode pH/ATC Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable
Connector TypesBNC (Waterproof), 8-pin Mini-DIN
JunctionCeramic
MaterialChemical Resistant Glass Body
pH Range0 to 14
Temperature (Metric) Operating0°C to 100°C
TypepH/ATC Combination Electrode
Filling Solution TypeROSS electrode 3 M KCl fill solution (Cat. No. 810007)
For Use With (Equipment)Orion Versa Star, Star A and Dual Star Meters
Internal ReferenceROSS
Isopotential Point7
pH Relative Accuracy0.03 pH for buffers using automatic temperature compensation
Precision0.01
Warranty2 Years
Length (Metric)120 mm
Length (Metric) Cable1 m
Diameter (Metric)12 mm
Unit SizeEach