accumet™ Mercury-free Conductivity Probes: Two Cell
Actual product may vary
accumet™ Mercury-free Conductivity Probes: Two Cell
accumet™ Mercury-free Conductivity Probes: Two Cell
accumet™ Mercury-free Conductivity Probes: Two Cell
accumet™ Mercury-free Conductivity Probes: Two Cell
accumet™ Mercury-free Conductivity Probes: Two Cell
accumet™ Mercury-free Conductivity Probes: Two Cell
For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCell ConstantMeasurement RangesMaterial
FIS13-620-1551 cm-110 to 2000 μS/cmGlass
FIS13-620-1601 cm-110 to 2000 μS/cmEpoxy
Catalog number FIS13-620-155
Price (SGD)
1,188.00
Each
Cell Constant:
1 cm-1
Measurement Ranges:
10 to 2000 μS/cm
Material:
Glass
Price (SGD)
1,188.00
Each
For use with AR60, AR50, AR20, AB30 benchtop and AP65 portable meters
Immersion-type cell's unique compact size is the same as a typical pH electrode—allowing use in standard electrode holder.
  • Two-cell types with glass or epoxy bodies
  • Dual pin connectors
  • Cell constants of 0.1, 1.0, or 10.0cm-1
Specifications
For Use With (Equipment)AR60, AR50, AR20, AB30 benchtop and AP65 portable meter
Cell Constant1 cm-1
TypeConductivity Probe
Connector Types2-pin
MaterialGlass
Measurement Ranges10 to 2000 μS/cm
Storage MethodImmersed in deionized water
Probe2 Cell Conductivity
Unit SizeEach