Autoclavable Bottletop Dispensers
Autoclavable Bottletop Dispensers
Actual product may vary
Autoclavable Bottletop Dispensers
Provides smooth, effortless dispensing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)IncrementsAccuracyPrecision
FIS13-615-1510 to 100 mL2.0 mL±0.6% (±3.000mL)±0.2% (±0.200mL)
FBD13615100.25 to 2.5 mL0.05 mL±0.6% (±0.015mL)±0.2% (±0.005mL)
FIS13-615-100.25 to 2.5 mL0.05 mL±0.6% (±0.015mL)±0.2% (±0.005mL)
FBD13615110.5 to 5 mL0.1 mL±0.6% (±0.030mL)±0.2% (±0.010mL)
FIS13-615-110.5 to 5 mL0.1 mL±0.6% (±0.030mL)±0.2% (±0.010mL)
FBD13615121 to 10 mL0.2 mL±0.6% (±0.060mL)±0.2% (±0.020mL)
FIS13-615-121 to 10 mL0.2 mL±0.6% (±0.060mL)±0.2% (±0.020mL)
FBD13615132.5 to 30 mL0.5 mL±0.6% (±0.180mL)±0.2% (±0.060mL)
FIS13-615-132.5 to 30 mL0.5 mL±0.6% (±0.180mL)±0.2% (±0.060mL)
FBD13615145 to 60 mL1.0 mL±0.6% (±0.360mL)±0.2% (±0.120mL)
FIS13-615-145 to 60 mL1.0 mL±0.6% (±0.360mL)±0.2% (±0.120mL)
FBD136151510 to 100 mL2.0 mL±0.6% (±3.000mL)±0.2% (±0.200mL)
Catalog number FIS13-615-15
Price (SGD)
985.00
EA
Add to cart
Volume (Metric):
10 to 100 mL
Increments:
2.0 mL
Accuracy:
±0.6% (±3.000mL)
Precision:
±0.2% (±0.200mL)
Price (SGD)
985.00
EA
Add to cart
Provides smooth, effortless dispensing
  • Wetted parts are PTFE, FEP and borosilicate glass (allows instrument to be used with a wide variety of chemicals)
  • 7 volume ranges to choose from
  • Bubble-free dispensing
  • Effortless plunger movement
  • Disassembles for easy cleaning and servicing
  • Autoclavable at 121°C, 15 psi for 10 to 15 minutes
  • Each bottle top dispenser includes one 28mm, 32mm, 38mm, 40mm, 45mm bottle neck adaptor
Component Description
  • Piston: PTFE
  • Cylinder: Glass
  • Volume Adjustment Knob: Screw Type
  • Valve Housing: PTFE
  • Valve Assembly: Glass Ball & Seal
  • Discharge Assembly: PTFE
  • Delivery Tube: FEP
  • Inlet Tube: FEP, 13 in. (with extension) 
  • Calibration: Individually calibrated and certified (in-lab by user is also possible)
  • Accuracy & Reproducibility: In accordance with ISO 8655 standards
  • Compability: Excellent with most reagents
Specifications
Accuracy±0.6% (±3.000mL)
AutoclavableAutoclavable
MaterialGlass
Volume (Metric)10 to 100 mL
Includes13-in. inlet tube (with extension)
Increments2.0 mL
Precision±0.2% (±0.200mL)
Unit SizeEA