Arsenal™ 5116 General Duty Gear Bag
Arsenal™ 5116 General Duty Gear Bag
Actual product may vary
Arsenal™ 5116 General Duty Gear Bag
Arsenal™ 5116 General Duty Gear Bag
Arsenal™ 5116 General Duty Gear Bag
Arsenal™ 5116 General Duty Gear Bag is designed to store, organize, and easily carry everything you need for the workday or weekend.
Have Questions?
Catalog NumberColor
FIS13-416Realtree Xtra™
Catalog number FIS13-416
Price (SGD)
2,212.00
CS
Add to cart
Color:
Realtree Xtra™
Price (SGD)
2,212.00
CS
Add to cart
  • Durable material withstands wear and tear
  • 1680D ballistic polyester base reinforces the bottom for ultimate durability
  • Water-resistant backing helps to repel water and stains
  • Heavy-duty webbing straps on top and side of duffel for extra reinforcement
  • Removable and adjustable padded shoulder strap for easy transportation
  • Heavy-duty zippers to secure gear inside the duffel
  • Nickel-plated Storm Drains™ added to each compartment for ventilation
  • Two large side pockets for cell phones, headphones, pens, keys, and other small parts
Specifications
ColorRealtree Xtra™
Material600D Polyester
Dimensions (L x W x H)23.5 x 9.5 x 12 in. (60 x 24 x 30 cm)
For Use With (Application)Contractors, Construction, Electricians, General Purpose
Quantity1 Ea.
SizeMedium
Volume (Metric)44 L
Unit SizeCS