Permanent Marking Pens
Permanent Marking Pens
Actual product may vary
Permanent Marking Pens
Use these Permanent Marking Pens to record results in lab notebooks, fill in charts, draw graphs, mark recording paper, and produce diagrams.
Have Questions?
Catalog NumberColor
FIS13-383DRed
Catalog number FIS13-383D
Price (SGD)
61.00
PK
Add to cart
Color:
Red
Price (SGD)
61.00
PK
Add to cart
Use these Permanent Marking Pens to record results in lab notebooks, fill in charts, draw graphs, mark recording paper, and produce diagrams.
  • Permanent marking pens with 0.3 mm tip.
  • Write on any smooth surface; ideal for plasticware, glassware, photos, slides, and more.
  • Waterproof, nonfading ink dries instantly to prevent smearing and bleeding.
  • Extra-fine, plastic writing tip stays sharp and precise.
  • Pen’s unique, long leg lays flat against rulers and flexible curves for accurate alignment.
  • Pens are perfectly balanced to comfortably fit in your hand.
  • Instant writing, long storage time, and dry-out prevention are ensured by an airtight ring seal in the cap.
  • Pen responds the moment it is touched to paper.
Specifications
TypePermanent Marking Pen
ColorRed
Quantity6 Pk.
Unit SizePK