Orion™ Conductivity Standards and Solutions
Actual product may vary
Orion™ Conductivity Standards and Solutions
Orion™ Conductivity Standards and Solutions
Thermo Scientific
Orion™ Conductivity Standards and Solutions
Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
Have Questions?
Catalog NumberPackagingConductivity
FIS13-301-135Bottle12.9 mS
Catalog number FIS13-301-135
Price (SGD)
465.00
Each
Packaging:
Bottle
Conductivity:
12.9 mS
Price (SGD)
465.00
Each
Ensure optimal conductivity probe maintenance and performance with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Conductivity conditioning solutions, conductivity standards and TDS standards are available for use with your Orion™ 2 Cell Conductivity Probe or Orion™ DuraProbe™ 4 Cell Lab and Field Conductivity Probes. Use these solutions during calibration, cleaning and storage of your probes to help eliminate build-up on the conductivity electrode cell or bulb surface and to attain accurate, consistent readings.
Conductivity Standards
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
  • For use with Thermo Scientific™ Orion™ Conductivity Probes
Warranty: Out of Box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionConductivity Standard
PackagingBottle
Traceability to NISTYes
Conductivity12.9 mS
Concentration0.1M KCl
ColorClear
pH4.75 to 7.75
Boiling Point100°C
For Use With (Equipment)Orion pH Meter
SolubilitySoluble
Solution TypeStandard Solution
TypeConductivity Standard
Quantity475 mL
Unit SizeEach