CHEMets Dissolved Oxygen Test Kit Refill
Actual product may vary
CHEMetrics
CHEMets Dissolved Oxygen Test Kit Refill
Provides results in two minutes or less
Have Questions?
Catalog NumberRangeTest MethodFor Use With (Equipment)
FIS13-299-202A0 to 40 ppb and 0 to 100 ppbRhodazine DC-7540 comparators
Catalog number FIS13-299-202A
Price (SGD)
166.00
Pack
Range:
0 to 40 ppb and 0 to 100 ppb
Test Method:
Rhodazine D
For Use With (Equipment):
C-7540 comparators
Price (SGD)
166.00
Pack
  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life
Specifications
AnalytesDissolved Oxygen (DO)
For Use With (Equipment)C-7540 comparators
Includes30 self-filling CHEMets ampoules to refill kits K-7540 (0-40 ppb) and K-7599 (0-100 ppb)
No. of Tests30
Range0 to 40 ppb and 0 to 100 ppb
Shelf Life1 year
Test MethodRhodazine D
Test Time2 min.
TypeCHEMets Dissolved Oxygen Refill
Unit SizePack