Polyseed™ BOD and Polyseed-NX™ CBOD Capsules
Actual product may vary
InterLab Supply
Polyseed™ BOD and Polyseed-NX™ CBOD Capsules
Produce fast, economical BOD analyses with consistent results. Available in two formulations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
FIS13-297-201Polyseed-NX CBOD Capsules
FIS13-297-200Polyseed BOD Capsules
Catalog number FIS13-297-201
Price (SGD)
537.00
Pack
Type:
Polyseed-NX CBOD Capsules
Price (SGD)
537.00
Pack
  • EPA accepted
  • Each capsule contains specialized microbial cultures for use in a broad range of areas involving degradation of industrial and municipal waste
  • To use, mix capsule contents with 0.5L of APHA standard dilution water at 20°C (68°F), stir for at least 60 minutes
  • Mixture yields seed inoculum for up to 200 BOD tests
  • For best results, use within one year of purchase
  • Also available as Polyseed-NX, which contains an approved chemical additive for the inhibition of ammonia nitrogen (NH3-N) in wastewater samples for CBOD analysis
Specifications
Certifications/ComplianceEPA
For Use With (Application)Standard seed source for the degradation of both industrial and municipal waste in CBOD testing
IncludesSpecialized microbial cultures
No. of TestsUp to 200
Shelf Life2 Years
TypePolyseed-NX CBOD Capsules
Unit SizePack