These plain form hydrometers are available in a variety of models measuring the relative densities (specific gravity) of oil and petroleum products in various ranges from -1° to 96°API in accordance with the American Petroleum Institute (API) and the American Society of Testing Materials (ASTM) E100 standards.
Have Questions?
Catalog Number
ASTM Number
Specific Gravity Range
Accuracy
Length (Metric)
FIS13-202-504
4H
29 to 41°
±0.1° API
330 mm
Catalog number FIS13-202-504
Price (SGD)
150.00
Each
ASTM Number:
4H
Specific Gravity Range:
29 to 41°
Accuracy:
±0.1° API
Length (Metric):
330 mm
Price (SGD)
150.00
Each
Plain form hydrometers measure the relative densities (specific gravity) of oil and petroleum products in various ranges from -1° to 96° API
Conform to the accuracy requirements of ASTM hydrometers
Lead free ballast and glass
Individually serialized
Temperature standardization: 60°F/60°F
Applications: Measures relative density (specific gravity) of oil and petroleum products.
Storage: ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.
Specifications
TypeHydrometer
ASTM Number4H
Accuracy±0.1° API
Increments0.1° API
Certifications/ComplianceTraceable to NIST
Length (Metric)330 mm
Ballast TypeLead Free
Baume Range29 to 41°
For Use With (Application)Measures relative density (specific gravity) of oil and petroleum products.
Specific Gravity Range29 to 41°
Unit SizeEach
Contents & Storage
ASTM hydrometers should be stored in a clean, dry, safe place. They should not be subjected to extreme temperatures.