Search Thermo Fisher Scientific
Catalog Number | Temperature Range (Metric) | Temperature Range (English) | Length (Metric) Overall |
---|---|---|---|
FIS13-201-641 | -10°C to +150°C | n.a. | 200 mm |
FIS13-201-521 | -100°C to +50°C | n.a. | 305 mm |
FIS13-201-555 | -10°C to +260°C | n.a. | 355 mm |
FIS13-201-640 | -10°C to +110°C | n.a. | 200 mm |
FIS13-201-643 | -35°C to +50°C | n.a. | 300 mm |
FIS13-201-644 | -20°C to +110°C | n.a. | 300 mm |
FIS13-201-645 | -20°C to +150°C | n.a. | 300 mm |
FIS13-201-646 | n.a. | -40°F to +120°F | 300 mm |
FIS13-201-647 | n.a. | 0°F to 230°F | 300 mm |
FIS13-201-649 | n.a. | 20°F to 500°F | 355 mm |
FIS13-201-648 | n.a. | 0°F to 300°F | 300 mm |