Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Mixers
▸
Mortars and Pestles
▸
Agate Mortar and Pestle Sets
Actual product may vary
Walter Stern
Agate Mortar and Pestle Sets
Ideal for preparing laboratory samples and high purity powders.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Outer
Height (Metric)
FIS12-950AA
5.08cm
2.3cm
FIS12-950B
9.52cm
4cm
2 Options
Catalog number
FIS12-950AA
Price (SGD)
297.00
Each
Add to cart
Diameter (Metric) Outer:
5.08cm
Height (Metric):
2.3cm
Price (SGD)
297.00
Each
Add to cart
Ideal for preparing laboratory samples and high purity powders.
Octagonally shaped, thick walled, agate mortar with pestle
Both have highly polished grinding surface
Specifications
Diameter (English) Outer
2 in.
Diameter (Metric) Outer
5.08cm
Product Type
Mortar and Pestle
Material
Agate Stone
For Use With (Application)
For grinding or crushing
Height (English)
0.9 in.
Height (Metric)
2.3cm
Finish
Highly Polished Grinding Surface