Osmette™ Micro-Osmette Osmometer
Actual product may vary
Precision Systems
Osmette™ Micro-Osmette Osmometer
Automatic digital osmometers measure osmolality in mOsm/kg H2O. Osmette Micro-Osmette Osmometer uses the freezing point depression method to provide a quick, accurate indication of total concentration. Instruments measure freezing points of solutions to 0.001°C, then directly translate them to measure of concentration.
Have Questions?
Catalog NumberVoltage
FIS12-827-7115 V
Catalog number FIS12-827-7
Price (SGD)
38,982.00
Each
Voltage:
115 V
Price (SGD)
38,982.00
Each
Easy to Use
  • Add sample tube to well, lower operating head to insert wire and thermistor temperature probe into the sample
  • Freezing point is detected by the probe, with accuracy of ±0.001°C
  • Digital display shows results directly in mOsm/kg H2O
  • Entire cycle takes between one-half and one minute per sample
Speedy and convenient
  • Fast cooldown, completely zero-stable meter
  • Reach operating temperature in <15 min. vs. 40 to 60 min. for other cryoscopic devices
  • Eliminate Dewar flasks, ice baths, calculations
  • Generate precise results in just 50 to 70 sec.; process 40 to 60 samples per hour
  • Minimal training; simple two-step calibration takes less than five minutes
  • All models hold calibration, eliminating daily calibration
Micro-Osmette, Model 5004
  • Automatic Operation
  • Features electronic cooling instead of a bath
  • Food and beverage industries, chemical manufacturing, pharmaceutical labs, research and quality control
  • Clinical applications include diagnosis of renal activity, disease electrolyte imbalance, hyper- and hyponatremia, measuring osmolality of body fluids
Specifications
Display TypeDigital
Frequency50/60 Hz
Voltage115 V
Cooling AgentThermoelectric Cooling Block
QuantityEach
Product TypeMicro Osmometer
Model5004
For Use With (Application)Food and beverage industries, chemical manufacturing, pharmaceutical labs, research and quality control, clinical applications include diagnosis of renal activity, disease electrolyte imbalance, hyper- and hyponatremia, measuring osmolality of body fluids
Height (English)10 in.
Height (Metric)25 cm
Length (English)15 in.
Measurement Time<1 min.
Length (Metric)38 cm
OperationAutomatic
Precision<1000mOsm/kg H2O: ±2mOsm/kg H2O, >1000mOsm/kg H2O: ±0.5%CV at 300mOsm/kg H2O: <1%
Range0 to 3000 mOsm/kg H2O
Sample Size50 μL
Width (Metric)13 cm
Width (English)5 in.
Cool-Down Time∼3 min.
Dimensions (L x W x H)15 x 5 x 10 in. (38 x 13 x 25 cm)
Electrical Requirements115 V 50/60 Hz
Unit SizeEach