Bath Fluid for Osmette™ Osmometers
Actual product may vary
Precision Systems
Bath Fluid for Osmette™ Osmometers
For use with Osmette A Osmometer, Osmette XL Osmometer.
Have Questions?
Catalog NumberVolume (Metric)
FIS12-827-22250 mL
Catalog number FIS12-827-22
Price (SGD)
117.00
Each
Volume (Metric):
250 mL
Price (SGD)
117.00
Each
  • Quantity: 250mL bottle
Specifications
Product TypeOsmometer Bath Fluid
For Use With (Equipment)12-8275, -3 and -33
Volume (English)8.45 oz.
Volume (Metric)250 mL
Unit SizeEach