Kimble™ KIMAX™ Babcock Bottle for Half & Half and Ice Cream testing
Kimble™ KIMAX™ Babcock Bottle for Half & Half and Ice Cream testing
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Babcock Bottle for Half & Half and Ice Cream testing
Size: 20% (2mL)
Have Questions?
Catalog NumberCapacity (Metric)
FIS12-725-302 mL
Catalog number FIS12-725-30
Price (SGD)
1,173.00
Case
Add to cart
Capacity (Metric):
2 mL
Price (SGD)
1,173.00
Case
Add to cart
  • Subdivisions: 0.2%
  • Tolerance: ±0.2%
  • Sample size: 9 grams
Specifications
Capacity (Metric)2 mL
MaterialBorosilicate Glass
Tolerance±0.2%
Height (Metric)165 mm
Product TypeBabcock Bottle
Height (English)6.5 in.
Unit SizeCase