Kimble™ KIMAX™ Babcock Bottles for Ice Cream Testing
Kimble™ KIMAX™ Babcock Bottles for Ice Cream Testing
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Babcock Bottles for Ice Cream Testing
Scale is marked with permanent brown stain on neck.
Have Questions?
Catalog NumberCapacity (Metric)
FIS12-700-152 mL
Catalog number FIS12-700-15
Price (SGD)
1,075.00
CS
Add to cart
Capacity (Metric):
2 mL
Price (SGD)
1,075.00
CS
Add to cart
  • Graduated to 10% in 0.1% subdivisions
  • Sample size: 18g
  • Overall height: 165mm
Specifications
Capacity (Metric)2 mL
MaterialGlass
Tolerance±0.1%
Height (Metric)165 mm
Product TypeBabcock Bottle
Height (English)6.5 in.
Unit SizeCS