Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Actual product may vary
Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Chill-Its™ 6660 Hard Hat Brim with Shade
Hard Hat Brim with Shade fits around the outside of a hard hat to provide sun and UV protection on the face and neck.
Have Questions?
Catalog NumberColor
FIS12-640Lime
Catalog number FIS12-640
Price (SGD)
55.00
PK
Add to cart
Color:
Lime
Price (SGD)
55.00
PK
Add to cart
  • 100% Polyester fabric
  • Universal design
  • Elastic inner rim to fit most hard hats
  • Designed to block sun and keep workers comfortable in outdoor environments
Specifications
ColorLime
For Use With (Application)Construction, Service, Landscaping, Material Handling
MaterialPolyester
Unit SizePK