Externally and Internally Threaded Cryogenic Storage Vials
Externally and Internally Threaded Cryogenic Storage Vials
Actual product may vary
Externally and Internally Threaded Cryogenic Storage Vials
Externally and Internally Threaded Cryogenic Storage Vials
Externally and Internally Threaded Cryogenic Storage Vials
Externally and Internally Threaded Cryogenic Storage Vials
Choice of externally threaded vials with HDPE closures for aseptic technique, or internally threaded vials featuring polypropylene closures with silicone gasket for tight seal
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Thread StyleClosure Material
FIS12-567-5001.2 mLInternalPolypropylene
FIS10-500-251.2 mLExternalHDPE
FIS10-500-262 mLExternalHDPE
FKS117879392 mLExternalHDPE
FIS10-500-275 mLExternalHDPE
FIS12-567-5012 mLInternalPolypropylene
FKS113216752 mLInternalPolypropylene
FIS12-567-5025 mLInternalPolypropylene
Catalog number FIS12-567-500
Price (SGD)
132.00
Pack
Add to cart
Volume (Metric):
1.2 mL
Thread Style:
Internal
Closure Material:
Polypropylene
Price (SGD)
132.00
Pack
Add to cart
Choice of externally threaded vials with HDPE closures for aseptic technique, or internally threaded vials featuring polypropylene closures with silicone gasket for tight seal
  • Sterile; not autoclavable
  • Noncytotoxic, nonpyrogenic
  • Large white writing area
  • Graduations in 0.5mL increments
  • Use with Fisherbrand Color Coders for easier sample identification
Externally threaded
  • Conical-style bottom
  • High density polyethylene
  • Self-standing
Internally threaded
  • With silicone gasket
  • Polypropylene
  • Conical- or round-style bottoms with a star-style foot
Vials are for research use only; not for in vitro diagnosis or parenterals.
Suitable for storage of biological materials in mechanical freezers or in the vapor phase of liquid nitrogen
Specifications
Diameter (English) Outer0.49 in.
ColorWhite
Diameter (Metric) Outer12.5 mm
MaterialPolypropylene
Pre-slitNo
SterilitySterile
Volume (Metric)1.2 mL
For Use With (Application)For storage of biological materials in mechanical freezers or in the vapor phase of liquid Nitrogen
Height (English)1.65 in.
No. per Case10
No. per Pack100/Pk.
StyleConical
Thread StyleInternal
Volume (Metric) Total1.2 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap
Quantity100 Pk., 10 Cs.
Marking SpotYes
For Use With (Equipment)Mechanical Freezers or LN Dewars
AutoclavableNot Autoclavable
Certifications/ComplianceNoncytotoxic, Nonpyrogenic
Height (Metric)42 mm
PreservativeNone
Product TypeCryogenic Vial
Unit SizePack