Search Fisher Scientific
| Catalog Number | Color | Holds | Material |
|---|---|---|---|
| FIS12-565-182 | Brown | 81 (1 to 1.8 mL Vials) | Cardboard |
| 330821 | White | 64 (1.5 mL Microcentrifuge Tubes) | Polycarbonate |
| FIS12-565-227 | White | 64 (1.5 mL Microcentrifuge Tubes) | Polycarbonate |
| FIS12-567-04 | White | 100 (3.6 mL Vials) | Cardboard |
| 341483 | White | 100 (3.6 mL Vials) | Polycarbonate |
| FIS12-565-234 | White | 100 (3.6 mL Vials) | Polycarbonate |
| NNU341483 | White | 100 (3.6 mL Vials) | Polycarbonate |
| 369639 | White | 81 (1 to 1.8 mL Vials) | Cardboard |
| FIS12-567-02 | White | 81 (3.6 mL Vials) | Cardboard |
| 374187 | Gray | 100 (1 to 1.8 mL Vials) | Polycarbonate |
| 5025-0505 | White | 25 (1.2 and 2 mL Vials) | Polycarbonate |
| FIS15-350-107 | White | 25 (1.2 and 2 mL Vials) | Polycarbonate |
| FKS10739331 | White | 25 (1.2 and 2 mL Vials) | Polycarbonate |
| 5026-0909 | White | 81 (1.2 and 2 mL Vials) | Polycarbonate |
| FIS15-350-107B | White | 81 (1.2 and 2 mL Vials) | Polycarbonate |
| 5026-1010 | Blue | 100 (1 to 1.5 mL Vials) | Polycarbonate |
| FIS03-337-7AA | Blue | 100 (1 to 1.5 mL Vials) | Polycarbonate |
| FIS03-337-7J | White | 81 (5 mL Vials) | Polycarbonate |
| 867013-0242 | Green | 81 (1 to 2 mL Vials) | Polycarbonate |
| 867013-0243 | Blue | 81 (1 to 2 mL Vials) | Polycarbonate |
| 867013-0245 | Assorted | 81 (1 to 2 mL Vials) | Polycarbonate |
| FIS14-175-65 | Assorted | 81 (1 to 2 mL Vials) | Polycarbonate |
| FKS10151042 | Assorted | 81 (1 to 2 mL Vials) | Polycarbonate |
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes. Choose from a variety of durable plastic Thermo Scientific™ CryoBox™ styles with keyed lids and printed location grids, or select economical cardboard/chipboard CryoBoxes™ for short-term storage. The plastic boxes are optimized to fit our own brand of Nalgene™ and Nunc™ vials and microcentrifuge tubes best.