Search Thermo Fisher Scientific
| Catalog Number | Culture Area | Closure Type |
|---|---|---|
| FIS12-556-010 | 75 cm² | Vented |
| FKS11830765 | 25 cm² | Vented |
| FKS11884235 | 75 cm² | Vented |
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated flasks with filter cap and seal cap options suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.