Premium Cover Glasses
Premium Cover Glasses
Actual product may vary
Premium Cover Glasses
The ultimate in purity and cleanliness for exacting work in histology, cytology, urinalysis and microbiology
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Quantity
FIS12-548B22 x 22 mmPack of 1 oz.
FIS12-544-1450 x 24 mmBulk Pack of 4 oz.
FIS12-544-1240 x 24 mmBulk Pack of 4 oz.
FIS12-548-5J40 x 24 mmPack of 1 oz.
FIS12-548-5E50 x 22 mmPack of 1 oz.
FIS12-548-5C40 x 22 mmPack of 1 oz.
FIS12-548-5R50 x 35 mmPack of 1 oz.
Catalog number FIS12-548B
Price (SGD)
-
Dimensions (L x W):
22 x 22 mm
Quantity:
Pack of 1 oz.
For exacting work in histology, cytology, urinalysis and microbiology. Experience the ultimate in purity and cleanliness.
  • Made of crystal-clear, pure water-white glass with uniform surface quality, precise squared edge, perfect flatness and exact dimensions
  • Precleaned and packaged to stay clean
  • Each glass is individually inspected and boxed in a molded plastic container for snugness and easy removal
Specifications
Product TypePremium Coverslip
ShapeSquare
MaterialGlass
Length (Metric)22 mm
Width (Metric)22 mm
Thickness (Metric)#1 (0.13 to 0.16 mm)
Dimensions (L x W)22 x 22 mm
QuantityPack of 1 oz.
Unit SizePK