Cover Glasses: Squares, Various Thicknesses
Cover Glasses: Squares, Various Thicknesses
Actual product may vary
Cover Glasses: Squares, Various Thicknesses
Made of finest optical borosilicate glass, with uniform thickness and size
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThickness (Metric)Length (Metric)Width (Metric)
FIS12-542C#1 (0.13 to 0.16 mm)25 mm25 mm
FIS12-541A#1.5 (0.16 to 0.19 mm)18 mm18 mm
FIS12-540A#2 (0.17 to 0.25 mm)18 mm18 mm
Catalog number FIS12-542C
Price (SGD)
-
Thickness (Metric):
#1 (0.13 to 0.16 mm)
Length (Metric):
25 mm
Width (Metric):
25 mm
Made of finest optical borosilicate glass, with uniform thickness and size
Square shape.
  • Corrosion-resistant
  • Lint-free and ready to use
  • Available in squares, circles and rectangles
  • Sold by weight; comes in a pack of 1 oz.
Specifications
Product TypeCoverslip
ShapeSquare
MaterialBorosilicate Glass
Length (Metric)25 mm
Width (Metric)25 mm
Thickness (Metric)#1 (0.13 to 0.16 mm)
For Use With (Application)Microscope, histology, cytology, hematology, microbiology
Quantity1 oz./Pk.
Unit SizePK