Hemacytometer Cover Glass is for your red blood cell and platelet counting chambers. Clear, non-fogging, non-corrosive glass. The chemical-neutral glass will not disturb the contents of the slide.
Have Questions?
Catalog Number
Quantity
FIS12-519-10
12 Pk.
Catalog number FIS12-519-10
Price (SGD)
115.00
Pack
Quantity:
12 Pk.
Price (SGD)
115.00
Pack
Thickness 0.4mm
Hemacytometer Cover Glass precisely fits the distance between the cover glass and the Hemacytometer counting chamber.
Made of clear, non-fogging, non-corrosive, chemically neutral glass.
Cleaned and washed.
Highly resistant to breakage.
Hemacytometer Cover Glass serves as a ready-to-use companion with the modern-day hemocytometer double-chamber format of the Blood Counting Chamber.
The Hemacytometer Cover Glass and Blood Counting Chamber has been essential tool for medical practitioners and biologists.
Specifications
Product TypeHemacytometer Coverslip
ShapeRectangle
MaterialGlass
Length (Metric)20 mm
Width (Metric)26 mm
Thickness (Metric)0.4 mm
Dimensions (L x W)20 x 26 mm
For Use With (Equipment)Double-ruled counting chamber