▸
Surgical and Dissection Equipment
▸
Dissection Needles
▸
Mid Grade Derf Needle Holders
Actual product may vary
Integra Miltex
Mid Grade Derf Needle Holders
Length 4.75 in. (12cm)
Have Questions?
Catalog Number
Length (English)
FIS12-460-836
4.75 in.
Catalog number
FIS12-460-836
Price (SGD)
71.00
Each
Add to cart
Length (English):
4.75 in.
Price (SGD)
71.00
Each
Add to cart
Specifications
Product Type
Derf Needle Holder
Material
Stainless Steel
Style
Serrated
Length (English)
4.75 in.
Length (Metric)
12 cm