MeisterHand™ Metzenbaum Scissors
Actual product may vary
Integra Miltex
MeisterHand™ Metzenbaum Scissors
Rounded tips for dissection
Have Questions?
Catalog NumberLength (Metric)Blade Style
FIS12-460-64817.8 cmStraight
Catalog number FIS12-460-648
Price (SGD)
179.00
Each
Length (Metric):
17.8 cm
Blade Style:
Straight
Price (SGD)
179.00
Each
  • Stainless steel
  • Cuts tough or delicate tissue
  • Non-glare matte finish
  • Straight or curved blades
  • Blunt points
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to help prevent tissue slippage
  • Black ring handles
Specifications
Blade StyleStraight
Length (English)7 in.
Length (Metric)17.8 cm
For Use With (Application)Surgical, Metzenbaum
Tip StyleBlunt/Blunt
Unit SizeEach