MeisterHand™ Allis Tissue Forceps
Actual product may vary
Integra Miltex
MeisterHand™ Allis Tissue Forceps
  • Stainless steel
  • Non-glare matte finish
Have Questions?
Catalog NumberLength (Metric)Teeth Count
FIS12-460-591146 mm4 X 5
Catalog number FIS12-460-591
Price (SGD)
319.00
Each
Length (Metric):
146 mm
Teeth Count:
4 X 5
Price (SGD)
319.00
Each
Specifications
Length (English)5.75 in.
MaterialStainless Steel
Length (Metric)146 mm
Product TypeForceps, Tissue, Allis
Quantity (No. per Case)Each
FinishNon-glare Matte
Teeth Count4 X 5
Unit SizeEach