Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Scissors
▸
Plastic Surgery Scissors
Actual product may vary
Integra Miltex
Plastic Surgery Scissors
For general-purpose plastic surgery
Have Questions?
Catalog Number
Length (Metric)
Blade Style
FIS12-460-484
12 cm
Straight
Catalog number
FIS12-460-484
Price (SGD)
329.00
Each
Add to cart
Length (Metric):
12 cm
Blade Style:
Straight
Price (SGD)
329.00
Each
Add to cart
High-quality surgical-grade stainless steel construction provides optimal performance
Fully tempered precision screw locks maintain perfect alignment
Ergonomic design combines weight, balance and feel for optimal comfort and performance
Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
Longer and slightly heavier than Iris scissors
Straight and curved patterns
Specifications
Product Type
Surgical Scissors
Blade Style
Straight
Grade
Premium
Length (English)
4.75 in.
Length (Metric)
12 cm
Material
Stainless Steel
For Use With (Application)
Surgical, Precision
Tip Style
Sharp/Blunt