Mayo Scissors
Actual product may vary
Integra Miltex
Mayo Scissors
Offer outstanding distal cutting capability
Have Questions?
Catalog NumberLength (English)Blade StyleProduct Type
FIS12-460-4729 in.Straight, BeveledMayo Scissors
Catalog number FIS12-460-472
Price (SGD)
557.00
Each
Length (English):
9 in.
Blade Style:
Straight, Beveled
Product Type:
Mayo Scissors
Price (SGD)
557.00
Each
  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Sharp/sharp scissors available in straight and curved patterns
  • Carb-N-Sert™ scissors have tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage
Specifications
Product TypeMayo Scissors
Blade StyleStraight, Beveled
GradePremium
Length (English)9 in.
Length (Metric)22.86 cm
MaterialStainless Steel
For Use With (Application)Dissecting
Unit SizeEach