▸
Surgical and Dissection Equipment
▸
Scalpels
▸
Sterile Standard Scalpels
Actual product may vary
Integra Miltex
Sterile Standard Scalpels
Stainless steel, razor-sharp cutting edge for superior cutting and clean incisions
Have Questions?
Catalog Number
Size
FIS12-460-451
# 10
Catalog number
FIS12-460-451
Price (SGD)
61.00
Pack
Add to cart
Size:
# 10
Price (SGD)
61.00
Pack
Add to cart
Sterile
Stainless steel, razor-sharp cutting edge
Safety sleeve for blade
Individually packaged
Sold in Pack of 10
Specifications
Blade Material
Stainless Steel
Product Type
Scalpel
For Use With (Application)
Superior Cutting and Clean Incisions
Size
# 10
Disposable
Single-use
Sterility
Sterile
Quantity
Pk. of 10
Shelf Life
5 Years