Instrument Trays
Actual product may vary
Integra Miltex
Instrument Trays
Manufactured using surgical grade, 304 stainless-steel
Have Questions?
Catalog NumberDimensions (L x W x H)
FIS12-460-43110 x 6.5 x 0.72 in.
Catalog number FIS12-460-431
Price (SGD)
76.00
Each
Dimensions (L x W x H):
10 x 6.5 x 0.72 in.
Price (SGD)
76.00
Each
  • Rolled edge
  • Thickness: 0.6mm
  • O.R. quality
  • Durable gauge to meet the needs of any healthcare customer
  • Available individually
Specifications
Dimensions (L x W x H)10 x 6.5 x 0.72 in.
MaterialStainless Steel
TypeInstrument Tray
Thickness0.6 mm
Unit SizeEach