Miltex™ Biopsy Punches with Plunger System
Actual product may vary
Integra
Miltex™ Biopsy Punches with Plunger System
Effortless ejection of lodged specimens.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FIS12-460-4104 mm
FIS12-460-4073 mm
FIS12-460-4052 mm
FIS12-460-4021 mm
FIS12-460-4031.5 mm
Catalog number FIS12-460-410
Price (SGD)
418.00
Pack
Diameter (Metric):
4 mm
Price (SGD)
418.00
Pack
Effortless ejection of lodged specimens
  • Seamless, stainless steel with razor-sharp cutting edge to eliminate torn or pulled biopsies
  • Ribbed handle provides comfort and control
  • Single-use
  • Sterile
  • Color coded with size clearly marked on each punch
  • 25/Pack
Specifications
Blade MaterialStainless Steel
Product TypeBiopsy Punch
For Use With (Application)Offer the added benefit of effortless ejection of Lodged Specimens
Handle TypeRibbed
DescriptionSeamless
DisposableSingle-use
SterilitySterile
IncludesPlunger System
Quantity25/Pk.
Shelf Life5 Years
Diameter (English)0.15 in.
Diameter (Metric)4 mm
Unit SizePack