▸
Surgical Tools
▸
Biopsy Punches
▸
Miltex™ Biopsy Punches with Plunger System
Actual product may vary
Integra
Miltex™ Biopsy Punches with Plunger System
Effortless ejection of lodged specimens.
Have Questions?
Change view
Catalog Number
Diameter (Metric)
FIS12-460-405
2 mm
FIS12-460-407
3 mm
FIS12-460-410
4 mm
FIS12-460-402
1 mm
FIS12-460-403
1.5 mm
5 Options
Catalog number
FIS12-460-405
Price (SGD)
418.00
Pack
Add to cart
Diameter (Metric):
2 mm
Price (SGD)
418.00
Pack
Add to cart
Effortless ejection of lodged specimens
Seamless, stainless steel with razor-sharp cutting edge to eliminate torn or pulled biopsies
Ribbed handle provides comfort and control
Single-use
Sterile
Color coded with size clearly marked on each punch
25/Pack
Specifications
Blade Material
Stainless Steel
Product Type
Biopsy Punch
For Use With (Application)
Offer the added benefit of effortless ejection of Lodged Specimens
Handle Type
Ribbed
Description
Seamless
Disposable
Single-use
Sterility
Sterile
Includes
Plunger System
Quantity
25/Pk.
Shelf Life
5 Years
Diameter (English)
0.079 in.
Diameter (Metric)
2 mm