Miltex™ Standard Biopsy Punches
Actual product may vary
Integra
Miltex™ Standard Biopsy Punches
Stainless-steel razor sharp cutting edge
Have Questions?
Catalog NumberDiameter (Metric)
FIS12-460-4000.15 cm
Catalog number FIS12-460-400
Price (SGD)
476.00
Pack
Diameter (Metric):
0.15 cm
Price (SGD)
476.00
Pack
Stainless-steel razor sharp cutting edge
  • Sealed in a sterile, peel-apart, see-through package
  • Sterilized by ethylene oxide
  • Single use
Specifications
Blade MaterialStainless Steel
Product TypeBiopsy Punch
For Use With (Application)Offer the Convenience of Single-use Disposability without Sacrificing quality, Feel or Performance
Handle TypeRibbed
DescriptionSeamless
DisposableSingle-use
SterilitySterile
Quantity50/Pk.
Shelf Life5 Years
Diameter (English)0.05 in.
Diameter (Metric)0.15 cm
Unit SizePack